Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
500 g
1 kg
Catalog number 404160025
Price (USD)/ Each
111.65
Online exclusive
124.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)/ Each
111.65
Online exclusive
124.00 
Save 12.35 (10%)
Add to cart
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Catalog number404160025
Price (USD)/ Each
111.65
Online exclusive
124.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS8001-79-4
IUPAC Name1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
Molecular FormulaC57H104O9
InChI KeyZEMPKEQAKRGZGQ-UHFFFAOYNA-N
SMILESCCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Refractive index1.4770 to 1.4810 (20°C, 589 nm)
Acid value=<2 mg KOH/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only