Thermo Scientific Chemicals

Cellulose Acetate Butyrate, Butyryl Content 50-54%, Thermo Scientific Chemicals

Catalog number: 404220025
2.5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Cellulose Acetate Butyrate, Butyryl Content 50-54%, Thermo Scientific Chemicals

Catalog number: 404220025
2.5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 404220025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9004-36-8
Viscosity
0.01 to 0.03 s
Appearance (Color)
White
Appearance (Form)
Crystalline or granular powder
Acetyl content
=<4 %
Hydroxyl content
1.5 % (typical)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text