Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
View all
Glass Bottle
Catalog number 404410025
Price (USD)/ Each
87.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
103.00 
Save 15.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
87.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
103.00 
Save 15.35 (15%)
Add to cart
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Catalog number404410025
Price (USD)/ Each
87.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
103.00 
Save 15.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium 2-chloroacetate
Molecular FormulaC2H2ClNaO2
InChI KeyFDRCDNZGSXJAFP-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)CCl
Molecular Weight (g/mol)116.48
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or granules or flakes
Titration with HClO497.5 to 102.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only