Thermo Scientific Chemicals

chloroacetic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 404421000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

chloroacetic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 404421000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 404421000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
541-88-8
IUPAC Name
2-chloroacetyl 2-chloroacetate
Molecular Formula
C4H4Cl2O3
InChI Key
PNVPNXKRAUBJGW-UHFFFAOYSA-N
SMILES
ClCC(=O)OC(=O)CCl
GC
>=96.0 %
Appearance (Form)
Powder or crystals and/or chunks or fused solid
Melting point
50°C to 60°C
Appearance (Color)
White to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text