Chlorobenzene, 99.6%, ACS reagent, Thermo Scientific Chemicals
Chlorobenzene, 99.6%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chlorobenzene, 99.6%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
View all
Glass Bottle
Catalog number 404490025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS108-90-7
IUPAC Namechlorobenzene
Molecular FormulaC6H5Cl
InChI KeyMVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5230 to 1.5250 (20°C, 589 nm)
Residue after evaporation=<0.02 %
Appearance (Form)Clear liquid
GC>=99.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only