Thermo Scientific Chemicals

chlorodifluoroacetic acid sodium salt, 96%, Thermo Scientific Chemicals

Catalog number: 404590250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

chlorodifluoroacetic acid sodium salt, 96%, Thermo Scientific Chemicals

Catalog number: 404590250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 404590250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1895-39-2
IUPAC Name
sodium 2-chloro-2,2-difluoroacetate
Molecular Formula
C2ClF2NaO2
InChI Key
MRTAVLDNYYEJHK-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)C(F)(F)Cl
Infrared spectrum
Conforms
Titration with HClO4
>=95 %
Appearance (Form)
Powder
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text