2-Chloroethanesulfonyl chloride, 95%, Thermo Scientific Chemicals
2-Chloroethanesulfonyl chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloroethanesulfonyl chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 404610250
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
106.00 
Save 15.35 (14%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
106.00 
Save 15.35 (14%)
Add to cart
2-Chloroethanesulfonyl chloride, 95%, Thermo Scientific Chemicals
Catalog number404610250
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
106.00 
Save 15.35 (14%)
-
Add to cart
Chemical Identifiers
CAS1622-32-8
IUPAC Name2-chloroethane-1-sulfonyl chloride
Molecular FormulaC2H4Cl2O2S
InChI KeyVHCSBTPOPKFYIU-UHFFFAOYSA-N
SMILESClCCS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
Titration Argentometric>=94 % (ex Cl)
Refractive index1.4910 to 1.4940 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only