4-(Chlorosulfonyl)benzoic acid, 96%, Thermo Scientific Chemicals
4-(Chlorosulfonyl)benzoic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Chlorosulfonyl)benzoic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 405060250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10130-89-9
IUPAC Name4-(chlorosulfonyl)benzoic acid
Molecular FormulaC7H5ClO4S
InChI KeyPTCSSXYPZOFISK-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Powder or crystals
Titration excess NaOH>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only