Thermo Scientific Chemicals

3-(Chlorosulfonyl)benzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 405070250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-(Chlorosulfonyl)benzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 405070250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 405070250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4052-92-0
IUPAC Name
3-(chlorosulfonyl)benzoyl chloride
Molecular Formula
C7H4Cl2O3S
InChI Key
XWEBTVZIZWEJOO-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=CC(=C1)S(Cl)(=O)=O
Appearance (Form)
Liquid after melting
Assay
>=97.5 %
Infrared spectrum
Conforms
Refractive index
1.5780 to 1.5800 (20°C, 589 nm)
Appearance (Color)
Clear yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text