Thermo Scientific Chemicals

5-Chloro-2-thiophenecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 405080010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Chloro-2-thiophenecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 405080010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 405080010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7283-96-7
IUPAC Name
5-chlorothiophene-2-carbaldehyde
Molecular Formula
C5H3ClOS
InChI Key
VWYFITBWBRVBSW-UHFFFAOYSA-N
SMILES
ClC1=CC=C(S1)C=O
Appearance (Color)
Clear yellow to orange-red
Appearance
May darken on storage
GC
>=96.0 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text