Chromium(VI) oxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Chromium(VI) oxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium(VI) oxide, 98+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 405230010
Price (USD)/ Each
865.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1018.00 
Save 152.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
865.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1018.00 
Save 152.35 (15%)
Add to cart
Chromium(VI) oxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Catalog number405230010
Price (USD)/ Each
865.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1018.00 
Save 152.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrioxochromium
Molecular FormulaCrO3
InChI KeyWGLPBDUCMAPZCE-UHFFFAOYSA-N
SMILESO=[Cr](=O)=O
Molecular Weight (g/mol)99.99
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder, crystals or granules
Insoluble matter=<0.01 %
Titration Iodometric>=98.0 %
Iron (Fe)=<200 ppm
Barium (Ba)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only