Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals
Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 405310250
Price (USD)/ Each
109.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
129.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
109.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
129.00 
Save 19.35 (15%)
Add to cart
Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals
Catalog number405310250
Price (USD)/ Each
109.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
129.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10534-89-1
IUPAC Namecobalt(3+) hexaamine trichloride
Molecular FormulaCl3CoH18N6
InChI KeyJXBGZYGSWFSYFI-UHFFFAOYSA-K
SMILESN.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.5 %
Appearance (Color)Orange to orange-brown
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only