Thermo Scientific Chemicals

Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 405311000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Hexaamminecobalt(III) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 405311000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 405311000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10534-89-1
IUPAC Name
cobalt(3+) hexaamine trichloride
Molecular Formula
Cl3CoH18N6
InChI Key
JXBGZYGSWFSYFI-UHFFFAOYSA-K
SMILES
N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]
Titration Argentometric
>=98.5 %
Appearance (Color)
Orange to orange-brown
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text