Thermo Scientific Chemicals

Copper(II) acetate, 99%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 405370010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Copper(II) acetate, 99%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 405370010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 405370010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
142-71-2
Infrared spectrum
Conforms
Titration with Na2S2O3
>=98.5 %
Appearance (Color)
Green to blue
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text