Copper(II) acetate, 99%, pure, anhydrous, Thermo Scientific Chemicals
Copper(II) acetate, 99%, pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) acetate, 99%, pure, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 405370500
Price (USD)/ Each
115.65
Online exclusive
136.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
115.65
Online exclusive
136.00 
Save 20.35 (15%)
Add to cart
Copper(II) acetate, 99%, pure, anhydrous, Thermo Scientific Chemicals
Catalog number405370500
Price (USD)/ Each
115.65
Online exclusive
136.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS142-71-2
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with Na2S2O3>=98.5 %
Appearance (Color)Green to blue
Appearance (Form)Crystalline powder

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
RUO – Research Use Only