Thermo Scientific Chemicals

Copper(II) Trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 405420250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Copper(II) Trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 405420250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 405420250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
34946-82-2
IUPAC Name
copper(II) triflate
Molecular Formula
C2H2CuF6O4S2
InChI Key
TXWAGEFXPDWAQH-UHFFFAOYSA-N
SMILES
[Cu].FC(F)(F)S(=O)=O.FC(F)(F)S(=O)=O
Appearance (Color)
White to light grey to light blue
Appearance (Form)
Powder
Titration with Na2S2O3
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text