Chemical Identifiers
IUPAC Nameethyl 4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
Molecular FormulaC18H19NO4
InChI KeyVMJKUPWQKZFFCX-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O
Molecular Weight (g/mol)313.35
View more