Thermo Scientific Chemicals

Coumarin 314, 99%, pure, laser grade, Thermo Scientific Chemicals

Catalog number: 405625000
500 mg, Each
Thermo Scientific Chemicals

Coumarin 314, 99%, pure, laser grade, Thermo Scientific Chemicals

Catalog number: 405625000
500 mg, Each
Quantity
Catalog number: 405625000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55804-66-5
IUPAC Name
ethyl 4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
Molecular Formula
C18H19NO4
InChI Key
VMJKUPWQKZFFCX-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O
HPLC
>=98.5 %
Appearance (Form)
Crystalline powder
Molar absorption coefficient
>=45000 l/(mol.cm) (at Lambda max)
Infrared spectrum
Conforms
Appearance (Color)
Yellow to orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text