Chemical Identifiers
CAS55804-65-4
IUPAC Name4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
Molecular FormulaC16H14NO4
InChI KeyKCDCNGXPPGQERR-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O
View more