Thermo Scientific Chemicals

Coumarin 343, Laser Grade, Thermo Scientific Chemicals

Catalog number: 405685000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

Coumarin 343, Laser Grade, Thermo Scientific Chemicals

Catalog number: 405685000
500 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 405685000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55804-65-4
IUPAC Name
4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
Molecular Formula
C16H14NO4
InChI Key
KCDCNGXPPGQERR-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O
Infrared spectrum
Conforms
UV Lambda max
409 nm (in Basic Ethanol)
Molar absorption coefficient
19900 l/(mol.cm) (at Lambda max)
Fluorescence max
470 nm (in Basic Ethanol)
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text