p-Cresol, 99+%, pure, Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Cresol, 99+%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
100 g
500 g
Catalog number 405740025
Price (USD)/ Each
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS106-44-5
IUPAC Name4-methylphenol
Molecular FormulaC7H8O
InChI KeyIWDCLRJOBJJRNH-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
GC>=99.0 %
AppearanceMay darken on storage
Water=<0.05 %
Appearance (Form)Crystalline solid or liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only