Cresyl Violet acetate, pure, certified, Thermo Scientific Chemicals
Cresyl Violet acetate, pure, certified, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cresyl Violet acetate, pure, certified, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
50 g
Packaging:
Glass bottle
Catalog number 405760100
Price (USD)/ Each
156.65
Online exclusive
184.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
156.65
Online exclusive
184.00 
Save 27.35 (15%)
Add to cart
Cresyl Violet acetate, pure, certified, Thermo Scientific Chemicals
Catalog number405760100
Price (USD)/ Each
156.65
Online exclusive
184.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name8,12-diamino-10λ⁴-oxa-5-azatetraphen-10-ylium acetate
Molecular FormulaC18H15N3O3
InChI KeyXKOCOMKJPWEOHX-UHFFFAOYSA-M
SMILESCC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
Molecular Weight (g/mol)321.34
View more
SpecificationsSpecification SheetSpecification Sheet
Dye content>=65 %
Absorption Ratio P-15/P+150.94 to 1.2
Appearance (Color)Dark green to black
BSC CertificationPasses
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only