Crystal Violet, certified
Crystal Violet, certified
Crystal Violet, certified
Crystal Violet, certified
Thermo Scientific Chemicals

Crystal Violet, certified

CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 405831000
Price (USD)
72.65
Special offer
Online exclusive
Ends: 26-Sep-2025
89.70
Save 17.05 (19%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 26-Sep-2025
89.70
Save 17.05 (19%)
Each
Add to cart
Chemical Identifiers
CAS548-62-9
IUPAC Nametris[4-(dimethylamino)phenyl]methylium chloride
Molecular FormulaC25H30ClN3
InChI KeyZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Dye content>=88 %
BSC CertificationPasses
Infrared spectrumConforms
UV Lambda max589 to 594 nm
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Share catalog number, name or link