Thermo Scientific Chemicals

Copper(II) chloride dihydrate, 99%, extra pure, powder, Thermo Scientific Chemicals

Catalog number: 405840025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Copper(II) chloride dihydrate, 99%, extra pure, powder, Thermo Scientific Chemicals

Catalog number: 405840025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 405840025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10125-13-0
IUPAC Name
copper(2+) dihydrate dichloride
Molecular Formula
Cl2CuH4O2
InChI Key
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Cu++]
Nitrate (NO3)
=<200 ppm
Appearance (Color)
Blue-green to blue
Titration Iodometric
>=98.5 %
Appearance (Form)
Crystalline powder or needles
Sulfate (SO4)
=<1000 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text