Copper(II) chloride dihydrate, 99%, extra pure, powder
Copper(II) chloride dihydrate, 99%, extra pure, powder
Copper(II) chloride dihydrate, 99%, extra pure, powder
Copper(II) chloride dihydrate, 99%, extra pure, powder
Thermo Scientific Chemicals

Copper(II) chloride dihydrate, 99%, extra pure, powder

CAS: 10125-13-0 | Cl2CuH4O2 | 170.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
2.5 kg
Packaging:
Glass bottle
Catalog number 405845000
Price (EUR)
47,52
Online Exclusive
55,90
Save 8,38 (15%)
Each
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
47,52
Online Exclusive
55,90
Save 8,38 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Namecopper(2+) dihydrate dichloride
Molecular FormulaCl2CuH4O2
InChI KeyMPTQRFCYZCXJFQ-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Cu++]
Molecular Weight (g/mol)170.48
View more
SpecificationsSpecification SheetSpecification Sheet
Nitrate (NO3)=<200 ppm
Appearance (Form)Crystalline powder or needles
Sulfate (SO4)=<1000 ppm
Zinc (Zn)=<500 ppm
Appearance (Color)Blue-green to blue
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link