Thermo Scientific Chemicals

Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 405851000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 405851000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 405851000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19004-19-4
IUPAC Name
dicopper(2+) pentahydrate tetranitrate
Molecular Formula
Cu2H10N4O17
InChI Key
IZDJJEMZQZQQQQ-UHFFFAOYSA-N
SMILES
O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Calcium (Ca)
=<50 ppm
Lead (Pb)
=<10 ppm
Potassium (K)
=<50 ppm
Appearance (Color)
Blue
Titration Iodometric
98.0 to 102.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text