Copper(II) oxide, ACS reagent, powder, Thermo Scientific Chemicals
Copper(II) oxide, ACS reagent, powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) oxide, ACS reagent, powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
Packaging:
View all
Plastic bottle
Catalog number 405860010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1317-38-0
IUPAC Namecopper(2+) oxidandiide
Molecular FormulaCuO
InChI KeyKKCXRELNMOYFLS-UHFFFAOYSA-N
SMILES[O--].[Cu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Potassium (K)=<200 ppm
Appearance (Color)Black to brown-black
Titration Complexometric>=99.0 %
Nitrogen compounds (as N)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only