Thermo Scientific Chemicals

Cyanogen iodide, 98%, Thermo Scientific Chemicals

Catalog number: 405960050
5 g, Each
Thermo Scientific Chemicals

Cyanogen iodide, 98%, Thermo Scientific Chemicals

Catalog number: 405960050
5 g, Each
Quantity
Catalog number: 405960050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
506-78-5
IUPAC Name
carbononitridic iodide
Molecular Formula
CIN
InChI Key
WPBXOELOQKLBDF-UHFFFAOYSA-N
SMILES
IC#N
Infrared spectrum
Conforms
Appearance (Color)
Off-white to light brown
Appearance (Form)
Crystalline powder
Titration Iodometric
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text