Thermo Scientific Chemicals

β-Cyclodextrin, 98%, Thermo Scientific Chemicals

Catalog number: 406005000
500 g, Each
Thermo Scientific Chemicals

β-Cyclodextrin, 98%, Thermo Scientific Chemicals

Catalog number: 406005000
500 g, Each
Quantity
Catalog number: 406005000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7585-39-9
IUPAC Name
5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
Molecular Formula
C42H70O35
InChI Key
WHGYBXFWUBPSRW-UHFFFAOYNA-N
SMILES
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
Appearance (Form)
Crystalline powder
Specific optical rotation
+159° to +165° (20°C, 589 nm) (c=1.5, water)
Appearance (Color)
White
HPLC
>=97.5 %
Specific optical rotation
on dry substance

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text