Cyclohexane, 99+%, for spectroscopy ACS, Thermo Scientific Chemicals
Cyclohexane, 99+%, for spectroscopy ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexane, 99+%, for spectroscopy ACS, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number 406025000
Price (USD)/ Each
60.65
Online exclusive
67.20 
Save 6.55 (10%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
60.65
Online exclusive
67.20 
Save 6.55 (10%)
Add to cart
Cyclohexane, 99+%, for spectroscopy ACS, Thermo Scientific Chemicals
Catalog number406025000
Price (USD)/ Each
60.65
Online exclusive
67.20 
Save 6.55 (10%)
-
Add to cart
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.0 %
Water=<0.02 % (Coulometric)
UVat 210 nm A: =<1
UVat 240 nm A: =<0.05
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only