Cyclohexanone, 99+%, Thermo Scientific Chemicals
Cyclohexanone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexanone, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Catalog number 406090025
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
Add to cart
Cyclohexanone, 99+%, Thermo Scientific Chemicals
Catalog number406090025
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS108-94-1
IUPAC Namecyclohexanone
Molecular FormulaC6H10O
InChI KeyJHIVVAPYMSGYDF-UHFFFAOYSA-N
SMILESO=C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.0 %
Water=<0.05 %
Residue after evaporation=<0.05 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only