Thermo Scientific Chemicals

Decyl sodium sulfate, HPLC grade, Thermo Scientific Chemicals

Catalog number: 406220250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Decyl sodium sulfate, HPLC grade, Thermo Scientific Chemicals

Catalog number: 406220250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 406220250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-87-0
IUPAC Name
sodium decyl sulfate
Molecular Formula
C10H21NaO4S
InChI Key
XZTJQQLJJCXOLP-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCOS([O-])(=O)=O
Titration after Ion exchange
>=98.5 % (On dry substance)
UV
(0.005 M in water) 1 cm cell
UV
at 220 nm A: =<0.05
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ion pair chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text