Dextran, Thermo Scientific Chemicals
Dextran, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dextran, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number 406275000
Price (USD)/ Each
431.65
Online exclusive
479.00 
Save 47.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
431.65
Online exclusive
479.00 
Save 47.35 (10%)
Add to cart
Dextran, Thermo Scientific Chemicals
Catalog number406275000
Price (USD)/ Each
431.65
Online exclusive
479.00 
Save 47.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9004-54-0
IUPAC NameDextran
Molecular Formula(C6H9O5)n(C6H10O5)m
SMILESO[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
MDL NumberMFCD00130935
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<7.0 %
Viscosity0.22 to 0.27 (1 % at 37°C)
Average molecular weight60000 to 90000
Appearance (Color)White to off-white
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only