1x1 image pixel for data collection
Dextrin
Dextrin
Dextrin
Thermo Scientific Chemicals

Dextrin

CAS: 9004-53-9 | C18H32O16 | 504.438 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 406281000
Price (USD)
23.65
Special offer
Online exclusive
Ends: 26-Sep-2025
29.10
Save 5.45 (19%)
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
23.65
Special offer
Online exclusive
Ends: 26-Sep-2025
29.10
Save 5.45 (19%)
Each
Add to cart
Chemical Identifiers
CAS9004-53-9
IUPAC Name(3R,4S,5S,6R)-2-{[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC18H32O16
InChI KeyFYGDTMLNYKFZSV-MRCIVHHJSA-N
SMILESOC[C@H]1OC(O[C@H]2[C@H](O)[C@@H](O)C(O[C@H]3[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Sulfated ash=<0.5 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link