Diacetin, 50%, tech. grade, remainder triacetin and monoacetin, Thermo Scientific Chemicals
Diacetin, 50%, tech. grade, remainder triacetin and monoacetin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diacetin, 50%, tech. grade, remainder triacetin and monoacetin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
Packaging:
Glass bottle
Catalog number 406300050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS25395-31-7, 102-76-1, 26446-35-5
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4400 to 1.4480 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=45 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only