Thermo Scientific Chemicals

trans-1,4-Diaminocyclohexane, 99%, Thermo Scientific Chemicals

Catalog number: 406360250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

trans-1,4-Diaminocyclohexane, 99%, Thermo Scientific Chemicals

Catalog number: 406360250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 406360250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2615-25-0
IUPAC Name
cyclohexane-1,4-diamine
Molecular Formula
C6H14N2
InChI Key
VKIRRGRTJUUZHS-UHFFFAOYSA-N
SMILES
NC1CCC(N)CC1
Melting point
68°C to 73°C
Appearance (Color)
White to yellow to brown
Appearance (Form)
Crystals and/or chunks
Infrared spectrum
Conforms
GC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text