Thermo Scientific Chemicals

4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals

Catalog number: 406411000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals

Catalog number: 406411000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 406411000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
81-11-8
IUPAC Name
5-amino-2-[(1E)-2-(4-amino-2-sulfophenyl)ethenyl]benzene-1-sulfonic acid
Molecular Formula
C14H14N2O6S2
InChI Key
REJHVSOVQBJEBF-OWOJBTEDSA-N
SMILES
NC1=CC=C(\C=C\C2=CC=C(N)C=C2S(O)(=O)=O)C(=C1)S(O)(=O)=O
Appearance (Color)
Cream to yellow-brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text