Thermo Scientific Chemicals

Dibenzyldithiocarbamic acid zinc salt, 95%, Thermo Scientific Chemicals

Catalog number: 406440250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Dibenzyldithiocarbamic acid zinc salt, 95%, Thermo Scientific Chemicals

Catalog number: 406440250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 406440250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14726-36-4
IUPAC Name
zinc(2+) bis((dibenzylcarbamothioyl)sulfanide)
Molecular Formula
C30H28N2S4Zn
InChI Key
AUMBZPPBWALQRO-UHFFFAOYSA-L
SMILES
[Zn++].[S-]C(=S)N(CC1=CC=CC=C1)CC1=CC=CC=C1.[S-]C(=S)N(CC1=CC=CC=C1)CC1=CC=CC=C1
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
183°C to 188°C
Assay
>=94 % (ex Zn)
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text