Thermo Scientific Chemicals

1,1-Dibromoethane, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 406481000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,1-Dibromoethane, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 406481000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 406481000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
557-91-5
IUPAC Name
1,1-dibromoethane
Molecular Formula
C2H4Br2
InChI Key
APQIUTYORBAGEZ-UHFFFAOYSA-N
SMILES
CC(Br)Br
Appearance (Color)
Clear colorless to brown
Refractive index
1.5090 to 1.5120 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text