1,4-Dibromonaphthalene, 98+%, Thermo Scientific Chemicals
1,4-Dibromonaphthalene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Dibromonaphthalene, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 406500250
Price (USD)/ Each
305.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
305.00
Add to cart
1,4-Dibromonaphthalene, 98+%, Thermo Scientific Chemicals
Catalog number406500250
Price (USD)/ Each
305.00
-
Add to cart
Chemical Identifiers
CAS83-53-4
IUPAC Name1,4-dibromonaphthalene
Molecular FormulaC10H6Br2
InChI KeyIBGUDZMIAZLJNY-UHFFFAOYSA-N
SMILESBrC1=C2C=CC=CC2=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Melting point79°C to 83°C
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only