1,3-Dibromo-2-propanone, 70%, tech.
1,3-Dibromo-2-propanone, 70%, tech.
1,3-Dibromo-2-propanone, 70%, tech.
1,3-Dibromo-2-propanone, 70%, tech.
Thermo Scientific Chemicals

1,3-Dibromo-2-propanone, 70%, tech.

CAS: 816-39-7 | C3H4Br2O | 215.87 g/mol
Have Questions?
Catalog NumberQuantityPackaging
40655025025 gGlass bottle
Catalog number 406550250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS816-39-7
IUPAC Name1,3-dibromopropan-2-one
Molecular FormulaC3H4Br2O
InChI KeyLQQKDSXCDXHLLF-UHFFFAOYSA-N
SMILESBrCC(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to dark brown
Appearance (Form)Liquid
GC>=65 %
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0417603Certificate of AnalysisMay 17, 2025406550250
A0417603Certificate of AnalysisMar 18, 2024406550250
A0392993Certificate of AnalysisDec 14, 2022406550250
A0417603Certificate of AnalysisMar 22, 2022406550250
4 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection