Thermo Scientific Chemicals

Dibutyl phosphate, 97%, Thermo Scientific Chemicals

Catalog number: 406645000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Dibutyl phosphate, 97%, Thermo Scientific Chemicals

Catalog number: 406645000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 406645000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
107-66-4
IUPAC Name
dibutoxyphosphinic acid
Molecular Formula
C8H19O4P
InChI Key
JYFHYPJRHGVZDY-UHFFFAOYSA-N
SMILES
CCCCOP(O)(=O)OCCCC
Titration with NaOH
>=96.0 %
Water
=<0.3 %
Appearance (Form)
Clear liquid
Color scale
=<100 APHA
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text