Thermo Scientific Chemicals

Dibutyl sebacate, 93%, Thermo Scientific Chemicals

Catalog number: 406650025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Dibutyl sebacate, 93%, Thermo Scientific Chemicals

Catalog number: 406650025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 406650025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
109-43-3
IUPAC Name
1,10-dibutyl decanedioate
Molecular Formula
C18H34O4
InChI Key
PYGXAGIECVVIOZ-UHFFFAOYSA-N
SMILES
CCCCOC(=O)CCCCCCCCC(=O)OCCCC
Color scale
=<40 APHA
Infrared spectrum
Conforms
Refractive index
1.4400 to 1.4420 (20°C, 589 nm)
Appearance (Form)
Clear liquid
Water
=<0.1 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text