Thermo Scientific Chemicals

1,2-Dichloroethane, 99+%, for spectroscopy ACS, Thermo Scientific Chemicals

Catalog number: 406830025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

1,2-Dichloroethane, 99+%, for spectroscopy ACS, Thermo Scientific Chemicals

Catalog number: 406830025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 406830025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
107-06-2
IUPAC Name
1,2-dichloroethane
Molecular Formula
C2H4Cl2
InChI Key
WSLDOOZREJYCGB-UHFFFAOYSA-N
SMILES
ClCCCl
Color scale
=<10 APHA
Water
=<0.03 % (Coulometric)
UV
at 245 nm A: =<0.05
UV
at 255 nm A: =<0.01
Residue after evaporation
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text