Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, Ac, Thermo Scientific Chemicals
Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, Ac, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, Ac, Thermo Scientific Chemicals

Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, AcroSeal™.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 406910010
Price (USD)/ Each
64.65
Online exclusive
76.10 
Save 11.45 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
76.10 
Save 11.45 (15%)
Add to cart
Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, Ac, Thermo Scientific Chemicals
Catalog number406910010
Price (USD)/ Each
64.65
Online exclusive
76.10 
Save 11.45 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
Molecular Weight (g/mol)84.93
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.85 %
Alkalinity=<0.0002 meq/g
UVat 235 nm A: =<1.0
Residue after evaporation=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only