Dichloromethane, 99.6%, ACS reagent, stabilized with amylene
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene
Thermo Scientific Chemicals

Dichloromethane, 99.6%, ACS reagent, stabilized with amylene

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
4 L
500 mL
1 L
25 L
Packaging:
View all
Metal Drum
Catalog number 406920250
Price (USD)
787.65
Special offer
Online exclusive
Ends: 26-Sep-2025
927.00
Save 139.35 (15%)
Each
-
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)
787.65
Special offer
Online exclusive
Ends: 26-Sep-2025
927.00
Save 139.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.5 %
Residue after evaporation=<0.002 %
Titrable acid=<0.0003 meq/g
Stabilizer20 to 60 ppm amylene
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link