Thermo Scientific Chemicals

Didodecyldimethylammonium bromide, 99%, Thermo Scientific Chemicals

Catalog number: 407120250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Didodecyldimethylammonium bromide, 99%, Thermo Scientific Chemicals

Catalog number: 407120250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 407120250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3282-73-3
IUPAC Name
didodecyldimethylazanium bromide
Molecular Formula
C26H56BrN
InChI Key
XRWMGCFJVKDVMD-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC
Infrared spectrum
Conforms
Melting point
158°C to 163°C
Titration Argentometric
>=98.5 %
Appearance (Color)
White to off-white
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text