Thermo Scientific Chemicals

Diethyl adipate, 99%, Thermo Scientific Chemicals

Catalog number: 407175000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl adipate, 99%, Thermo Scientific Chemicals

Catalog number: 407175000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 407175000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
141-28-6
IUPAC Name
1,6-diethyl hexanedioate
Molecular Formula
C10H18O4
InChI Key
VIZORQUEIQEFRT-UHFFFAOYSA-N
SMILES
CCOC(=O)CCCCC(=O)OCC
GC
>=98.5 %
Refractive index
1.4260 to 1.4280 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text