Thermo Scientific Chemicals

Diethyl chlorophosphate, 95%, Thermo Scientific Chemicals

Catalog number: 407240250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl chlorophosphate, 95%, Thermo Scientific Chemicals

Catalog number: 407240250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 407240250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
814-49-3
IUPAC Name
diethyl phosphorochloridate
Molecular Formula
C4H10ClO3P
InChI Key
LGTLXDJOAJDFLR-UHFFFAOYSA-N
SMILES
CCOP(Cl)(=O)OCC
GC
>=94 %
Refractive index
1.4160 to 1.4180 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text