Diethyl chlorophosphate, 95%, Thermo Scientific Chemicals
Diethyl chlorophosphate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl chlorophosphate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 407241000
Price (USD)/ Each
92.65
Online exclusive
109.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
92.65
Online exclusive
109.00 
Save 16.35 (15%)
Add to cart
Diethyl chlorophosphate, 95%, Thermo Scientific Chemicals
Catalog number407241000
Price (USD)/ Each
92.65
Online exclusive
109.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namediethyl phosphorochloridate
Molecular FormulaC4H10ClO3P
InChI KeyLGTLXDJOAJDFLR-UHFFFAOYSA-N
SMILESCCOP(Cl)(=O)OCC
Molecular Weight (g/mol)172.54
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only